Merck
  • Oxalic Diamides and tert-Butoxide: Two Types of Ligands Enabling Practical Access to Alkyl Aryl Ethers via Cu-Catalyzed Coupling Reaction.
Oxalic Diamides and tert-Butoxide: Two Types of Ligands Enabling Practical Access to Alkyl Aryl Ethers via Cu-Catalyzed Coupling Reaction.
Journal of the American Chemical Society (2019-01-29)
Zhixiang Chen, Yongwen Jiang, Li Zhang, Yinlong Guo, Dawei Ma
ABSTRACT
A robust and practical protocol for preparing alkyl aryl ethers has been developed, which relies on using two types of ligands to promote Cu-catalyzed alkoxylation of (hetero)aryl halides. The reaction scope is very general for a variety of coupling partners, particularly for challenging secondary alcohols and (hetero)aryl chlorides. In case of coupling with aryl chlorides and bromides, two oxalic diamides serve as the powerful ligands. The tert-butoxide is first demonstrated as a ligand for Cu-catalyzed coupling reaction, leading to alkoxylation of aryl iodides complete at room temperature. Additionally, a number of carbohydrate derivatives are applicable for this coupling reaction, affording the corresponding carbohydrate-aryl ethers in 29-98% yields.
Sigma-Aldrich
BNMO, ≥95%
Sigma-Aldrich
DPEO, ≥95%