Merck
Crbn modulates calcium influx by regulating Orai1 during efferocytosis.
Nature communications (2020-11-01)
Hyunji Moon, Chanhyuk Min, Gayoung Kim, Deokhwan Kim, Kwanhyeong Kim, Sang-Ah Lee, Byeongjin Moon, Susumin Yang, Juyeon Lee, Seung-Joo Yang, Steve K Cho, Gwangrog Lee, Chang Sup Lee, Chul-Seung Park, Daeho Park
ABSTRACT
Calcium flux regulating intracellular calcium levels is essential and modulated for efficient efferocytosis. However, the molecular mechanism by which calcium flux is modulated during efferocytosis remains elusive. Here, we report that Orai1, a Crbn substrate, is upregulated via its attenuated interaction with Crbn during efferocytosis, which increases calcium influx into phagocytes and thereby promotes efferocytosis. We found that Crbn deficiency promoted phagocytosis of apoptotic cells, which resulted from facilitated phagocytic cup closure and was nullified by a CRAC channel inhibitor. In addition, Orai1 associated with Crbn, resulting in ubiquitination and proteasomal degradation of Orai1 and alteration of SOCE-mediated calcium influx. The association of Orai1 with Crbn was attenuated during efferocytosis, leading to reduced ubiquitination of Orai1 and consequently upregulation of Orai1 and calcium influx. Collectively, our study reveals a regulatory mechanism by which calcium influx is modulated by a Crbn-Orai1 axis to facilitate efferocytosis.
Sigma-Aldrich
9-Anthracenecarboxylic acid, 99%
Sigma-Aldrich
Mdivi-1, ≥98% (HPLC), powder
Sigma-Aldrich
YM-58483, ≥98% (HPLC)
Sigma-Aldrich
Anti-CRBN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Crbn
Sigma-Aldrich
MISSION® esiRNA, targeting human CRBN
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests