Merck
  • TARPs gamma-2 and gamma-7 are essential for AMPA receptor expression in the cerebellum.
TARPs gamma-2 and gamma-7 are essential for AMPA receptor expression in the cerebellum.
The European journal of neuroscience (2010-06-10)
Maya Yamazaki, Masahiro Fukaya, Kouichi Hashimoto, Miwako Yamasaki, Mika Tsujita, Makoto Itakura, Manabu Abe, Rie Natsume, Masami Takahashi, Masanobu Kano, Kenji Sakimura, Masahiko Watanabe
ABSTRACT
The alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA)-type glutamate receptors require auxiliary subunits termed transmembrane AMPA receptor regulatory proteins (TARPs), which promote receptor trafficking to the cell surface and synapses and modulate channel pharmacology and gating. Of six TARPs, gamma-2 and gamma-7 are the two major TARPs expressed in the cerebellum. In the present study, we pursued their roles in synaptic expression of cerebellar AMPA receptors. In the cerebellar cortex, gamma-2 and gamma-7 were preferentially localized at various asymmetrical synapses. Using quantitative Western blot and immunofluorescence, we found severe reductions in GluA2 and GluA3 and mild reduction in GluA4 in gamma-2-knockout (KO) cerebellum, whereas GluA1 and GluA4 were moderately reduced in gamma-7-KO cerebellum. GluA2, GluA3 and GluA4 were further reduced in gamma-2/gamma-7 double-KO (DKO) cerebellum. The large losses of GluA2 and GluA3 in gamma-2-KO mice and further reductions in DKO mice were confirmed at all asymmetrical synapses examined with postembedding immunogold. Most notably, the GluA2 level in the postsynaptic density fraction, GluA2 labeling density at parallel fiber-Purkinje cell synapses, and AMPA receptor-mediated currents at climbing fiber-Purkinje cell synapses were all reduced to approximately 10% of the wild-type levels in DKO mice. On the other hand, the reduction in GluA4 in gamma-7-KO granular layer reflected its loss at mossy fiber-granule cell synapses, whereas that of GluA1 and GluA4 in gamma-7-KO molecular layer was caused, at least partly, by their loss in Bergmann glia. Therefore, gamma-2 and gamma-7 cooperatively promote synaptic expression of cerebellar AMPA receptors, and the latter also promotes glial expression.
Sigma-Aldrich
Anti-Synaptophysin Antibody, clone SY38, clone SY38, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glutamate Receptor 2 Antibody, extracellular, clone 6C4, clone 6C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glutamate Receptor 3 Antibody, clone 3B3, clone 3B3, Chemicon®, from mouse