Merck
  • Nanogyroids incorporating multivalent lipids: enhanced membrane charge density and pore forming ability for gene silencing.
Nanogyroids incorporating multivalent lipids: enhanced membrane charge density and pore forming ability for gene silencing.
Langmuir : the ACS journal of surfaces and colloids (2011-05-27)
Cecília Leal, Kai K Ewert, Rahau S Shirazi, Nathan F Bouxsein, Cyrus R Safinya
ABSTRACT
The self-assembly of a custom-synthesized pentavalent cationic lipid (MVL5) and glycerol monooleate (GMO) with small interfering RNA (siRNA) results in the formation of a double-gyroid bicontinuous inverted cubic phase with colocalized lipid/siRNA domains as shown by synchrotron X-ray scattering and fluorescence microscopy. The high charge density (due to MVL5) and positive Gaussian modulus of the GMO-containing membranes confer optimal electrostatic and elastic properties for endosomal escape, enabling efficient siRNA delivery and effective, specific gene silencing.
Product Description
Avanti
MVL5, Avanti Research - A Croda Brand 890000P, powder