Merck
  • Oxeiptosis, a ROS-induced caspase-independent apoptosis-like cell-death pathway.
Oxeiptosis, a ROS-induced caspase-independent apoptosis-like cell-death pathway.
Nature immunology (2017-12-20)
Cathleen Holze, Chloé Michaudel, Claire Mackowiak, Darya A Haas, Christian Benda, Philipp Hubel, Friederike L Pennemann, Daniel Schnepf, Jennifer Wettmarshausen, Marianne Braun, Daisy W Leung, Gaya K Amarasinghe, Fabiana Perocchi, Peter Staeheli, Bernhard Ryffel, Andreas Pichlmair
ABSTRACT
Reactive oxygen species (ROS) are generated by virus-infected cells; however, the physiological importance of ROS generated under these conditions is unclear. Here we found that the inflammation and cell death induced by exposure of mice or cells to sources of ROS were not altered in the absence of canonical ROS-sensing pathways or known cell-death pathways. ROS-induced cell-death signaling involved interactions among the cellular ROS sensor and antioxidant factor KEAP1, the phosphatase PGAM5 and the proapoptotic factor AIFM1. Pgam5 -/- mice showed exacerbated lung inflammation and proinflammatory cytokines in an ozone-exposure model. Similarly, challenge with influenza A virus led to increased infiltration of the virus, lymphocytic bronchiolitis and reduced survival of Pgam5 -/- mice. This pathway, which we have called 'oxeiptosis', was a ROS-sensitive, caspase independent, non-inflammatory cell-death pathway and was important for protection against inflammation induced by ROS or ROS-generating agents such as viral pathogens.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-HA−Peroxidase antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-PGAM5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.