Merck
  • Multiple reaction monitoring targeted LC-MS analysis of potential cell death marker proteins for increased bioprocess control.
Multiple reaction monitoring targeted LC-MS analysis of potential cell death marker proteins for increased bioprocess control.
Analytical and bioanalytical chemistry (2018-04-03)
Simone Albrecht, Christian Kaisermayer, David Reinhart, Monica Ambrose, Renate Kunert, Anna Lindeberg, Jonathan Bones
ABSTRACT
The monitoring of protein biomarkers for the early prediction of cell stress and death is a valuable tool for process characterization and efficient biomanufacturing control. A representative set of six proteins, namely GPDH, PRDX1, LGALS1, CFL1, TAGLN2 and MDH, which were identified in a previous CHO-K1 cell death model using discovery LC-MSE was translated into a targeted liquid chromatography multiple reaction monitoring mass spectrometry (LC-MRM-MS) platform and verified. The universality of the markers was confirmed in a cell growth model for which three Chinese hamster ovary host cell lines (CHO-K1, CHO-S, CHO-DG44) were grown in batch culture in two different types of basal media. LC-MRM-MS was also applied to spent media (n = 39) from four perfusion biomanufacturing series. Stable isotope-labelled peptide analogues and a stable isotope-labelled monoclonal antibody were used for improved protein quantitation and simultaneous monitoring of the workflow reproducibility. Significant increases in protein concentrations were observed for all viability marker proteins upon increased dead cell numbers and allowed for discrimination of spent media with dead cell densities below and above 1 × 106 dead cells/mL which highlights the potential of the selected viability marker proteins in bioprocess control. Graphical abstract Overview of the LC-MRM-MS workflow for the determination of proteomic markers in conditioned media from the bioreactor that correlate with CHO cell death.
Sigma-Aldrich
SILuMAB K1 Stable-Isotope Labeled Universal Monoclonal Antibody, recombinant, expressed in CHO cells
Pricing and availability is not currently available.