
Vitexin 2′′-rhamnoside

Synonym(s):
Vitexin 2-O-rhamnoside, 2-O-Rhamnosylvitexin
Empirical Formula (Hill Notation):
C27H30O14
CAS Number:
Molecular Weight:
578.52
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing