Stannous chloride
Synonym(s):
Tin(II) chloride, Stannous chloride
Linear Formula:
SnCl2
CAS Number:
Molecular Weight:
189.62
Products
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Product Line
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing