Skip to Content
Merck
Salvianolic acid B analytical standard

Salvianolic acid B

Salvianolic acid B analytical standard
Synonym(s):
(2S,3S)-4-[(1E)-3-[(1R)-1-Carboxy-2-(3,4-dihydroxyphenyl)ethoxy]-3-oxo-1-propen-1-yl]-2-(3,4-dihydroxyphenyl)-2,3-dihydro-7-hydroxy-3-benzofurancarboxylic acid 3-[(1R)-1-carboxy-2-(3,4-dihydroxyphenyl)ethyl] ester, Dan Shen Suan B, Danfensuan B, Lithospermic acid B, Salvianolic acid B
Empirical Formula (Hill Notation):
C36H30O16
CAS Number:
Molecular Weight:
718.61

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique