
ORBO™ 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters

Synonym(s):
ORBO™ Activated Silica Gel Tube
Products
Brand
Product Line
Agency
Chemical Composition
OD (mm)
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing