![N,N,N′,N′-Tetramethyl-1,8-naphthalenediamine purum, ≥99.0% (NT)](/deepweb/assets/sigmaaldrich/product/structures/394/790/58d8bb05-a997-42f8-b745-ae734fffb2e6/640/58d8bb05-a997-42f8-b745-ae734fffb2e6.png)
N,N,N′,N′-Tetramethyl-1,8-naphthalenediamine
![N,N,N′,N′-Tetramethyl-1,8-naphthalenediamine purum, ≥99.0% (NT)](/deepweb/assets/sigmaaldrich/product/structures/394/790/58d8bb05-a997-42f8-b745-ae734fffb2e6/640/58d8bb05-a997-42f8-b745-ae734fffb2e6.png)
Synonym(s):
1,8-Bis(dimethylamino)naphthalene, DMAN, Proton-sponge®
Empirical Formula (Hill Notation):
C14H18N2
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing