
Bumetanide impurity B

Synonym(s):
3-Amino-4-phenoxy-5-sulphamoylbenzoic acid, Desbutylbumetanide
Empirical Formula (Hill Notation):
C13H12N2O5S
CAS Number:
Molecular Weight:
308.31
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing