Now, every lab bench can easily access freshly purified ultrapure water—even those without a direct water supply. New Milli-Q® SQ 2Series water purification systems are designed to be simple, robust and innovative. They're also fully scalable, allowing you to focus on what matters most: your science. These systems have quicky become a lab favorite, earning the title of 'Best New General Lab Product of 2024' in the SelectScience® Scientists' Choice Awards.
- How It Works
- System Configurations
- Features & Benefits
- Greener Alternative Products
- Related Videos
- Download Brochure
- Ordering Information
To view installation and demo videos or download user manuals, visit our Installation & Use Page.
How Milli-Q® SQ 2Series Lab Water Systems Work
Thanks to its completely scalable design, any lab bench with an electrical outlet can easily access fresh purified 18.2 MΩ∙cm ultrapure water.
- Rapidly fill the 3.5 L Switch tank with reverse osmosis (RO) water in as few as 6 minutes.
- Easily carry the Switch tank to your science.
- Dispense freshly polished 18.2 MΩ∙cm ultrapure water at any bench.
- Repeat as needed.
Milli-Q® SQ 2Series System Configurations
The Milli-Q® SQ 2Series offers a choice of configurations that make the system adaptable and scalable to nearly any lab or building setup. Simply select the setup that best fits your space and user requirements, then easily expand with more modules as your lab evolves. Features common to nearly all Milli-Q® SQ 2Series system configurations include:
- Rapid pure RO production at a rate of up to 42 L/h, filling the 3.5 L Switch tank in as few as 6 min
- Ergonomic dispensing of ultrapure water, from 1.6 L/min to drop by drop
- Simply expand capabilities by adding autonomous ultrapure water dispensing modules (ZSQ200U0T0)
Milli-Q® SQ 240 (ZSQ240R0TK)
- Standard configuration
- Suitable for one small team
- Install dispensing module next to an ultrapure water need
Milli-Q® SQ 240L (ZSQ240RLTK)
- Dual tank configuration
- Suitable for multiple or larger teams
- Install dispensing module next to an ultrapure water need
Milli-Q® SQ 240XL (ZSQ240RXTK)
- 50L tank configuration
- Suitable for direct feed of lab equipment from the 50 L tank
- Install dispensing module next to an ultrapure water need
Milli-Q® SQ 240C (ZSQ240UCTK)
- Combined configuration
- Suitable for one small lab or team
- Install system next to a sink
Milli-Q® SQ 240CV (ZSQ240UCVTK)
- Combined, preset volume configuration
- Suitable for routine lab activities
- Repeat serial ultrapure water preset volume dispense from a fixed point
- Install system next to a sink
Milli-Q® SQ 200P (ZSQ200UPT0)
- Pressurized loop configuration
- Connect system to a pressurized pure water loop or tank
- Expand capabilities by connecting additional systems to the pressurized pure water loop
Here are just some of the benefits you’ll experience with a Milli-Q® SQ 2Series system in your lab:
Ultimate Simplicity
The system is easy and intuitive to install, operate and maintain for any user.
- Proven, do-it-yourself self-installation (~30 min)
- Universal interface
- Ergonomic dispensing
- Mobile, transparent tank
- Twist & Lock cartridge changes
- No need to replace the UV lamp
Ultra Compact
The uniquely small size leaves space for your science at any lab bench.
- System units have the same footprint as a sheet of paper: 22x27 cm/unit
- Only an electrical outlet (and no water inlet) is needed to dispense ultrapure water
Unlimited Scalability
Find the configuration that best fits your space and user requirements. Then easily expand with unlimited ultrapure water dispensing modules as your lab evolves.
Reliable Quality
Proven Milli-Q® water purification technologies mean you can trust your system is always ready to dispense consistent quality 18.2 MΩ∙cm @25 oC ultrapure water.
- High efficiency removal of ions down to trace levels with SQPAK™ Quanta polishing cartridge
- For organic-sensitive applications, SQPAK™ TOC Quanta cartridge contains activated carbon
- Pure RO water quality is maintained by regular rinsing of the RO membrane
- Choice of final filters can match your specific application needs
Milli-Q® SQ 2Series Water Systems are Greener Alternative Products
As part of our Design for Sustainability (DfS) approach to product development, Milli-Q® SQ 2Series systems were developed to deliver high-quality ultrapure water at multiple locations with a reduced environmental footprint compared to previous generation Direct-Q® systems.
As a result of these efforts, these new systems are identified as Greener Alternative Products thanks to the following improvements versus previous generation systems:
- Consume less electricity thanks to simplified design, minimized electronics, and optimized stand-by mode (where most energy is consumed)
- Consume less water during manufacturing and use
- Limited material requirements if 2 or more dispensing locations thanks to a modular design
- Packaging consists of 100% recycled cardboard with sustainable forestry certification, and protective inserts made of PE with 50% recycled content
The precise sustainability gains depend on the selected Milli-Q® SQ 2Series system configuration. We invite you to review the DfS Scorecard for the below three configurations to support you in making greener purchasing decision:
| System Configuration | Water Purification Step | Download DfS Scorecard |
| Milli-Q® SR 240/240L/240XL | Tap to Pure RO Water | |
| Milli-Q® SQ 240/240L/240XL/240C/240CV | Tap to Ultrapure Water | |
| Milli-Q® SQ 200 | Pure RO to Ultrapure Water | Download PDF |
| Milli-Q® SQ 200P | Pure to Ultrapure Water |
- Learn more about how all new Milli-Q® lab water systems and processes are being designed to be more sustainable.
- Learn more about the sustainability criteria that Greener Alternative Products must fulfill.
Related Resources
Milli-Q® SQ System Selection Guide
Select and configure the Milli-Q® SQ system setup that best matches your space and user requirements.
- Installation & Use of Milli-Q® SQ 2Series Water Purification Systems
Be up and running in no time with instructional videos and user manuals for Milli-Q® SQ 2Series lab water systems.
- Milli-Q® SQ 2Series Systems
A robust system for your essential science.
- Lab Water Selection Guide
Identify and configure your optimal water purification system.
- Milli-Q® Ultrapure Water Solutions
Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.
- Milli-Q® IQ 7003/05/10/15 Ultrapure & Pure Lab Water Purification System
Milli-Q® IQ 7003/05/10/15 lab water systems provide consistent-quality Type 1 and Type 2 water, are intuitive to use, and can support your lab's sustainability goals.
- Milli-Q® IQ 7000 Ultrapure Water System
Discover an ultrapure lab water system designed with you in mind. Intelligent, intuitive and connected, the Milli-Q® IQ 7000 system will simplify the way you work in the lab.
Purification, Storage and Dispensing
Response not successful: Received status code 500
Purification Cartridges and Final Filters
Response not successful: Received status code 500
Accessories
Response not successful: Received status code 500
Power Cords
Response not successful: Received status code 500
Milli-Q® Services
Response not successful: Received status code 500