GC Analysis of Propylene Oxide Enantiomers on Astec® CHIRALDEX™ A-TA
Materials
analytical column
Product No.
Description
Pricing
standard
Product No.
Description
Pricing
CONDITIONS
column
Astec CHIRALDEX A-TA, 30 m x 0.25 mm I.D., 0.12 μm (73003AST)
oven
30 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 24 psi
injection
3 μL, 80:1 split
sample
peak 1: R(+)-propylene oxide
peak 2: S(-)-propylene oxide
Description
Analysis Note
Can also substitute CHIRALDEX B-DP for this separation.
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC