Merck
  • Modulation of the biomarkers of inflammation and oxidative stress by ruminant trans fatty acids and dairy proteins in vascular endothelial cells (HUVEC).
Modulation of the biomarkers of inflammation and oxidative stress by ruminant trans fatty acids and dairy proteins in vascular endothelial cells (HUVEC).
Prostaglandins, leukotrienes, and essential fatty acids (2017-10-17)
Marine S Da Silva, Jean-François Bilodeau, Jessica Larose, Karine Greffard, Pierre Julien, Olivier Barbier, Iwona Rudkowska
ABSTRACT
This study aimed to determine whether dairy macronutrients alter markers of inflammation and oxidative stress in endothelial cells. Human endothelial cells (HUVEC) were treated with ruminant trans fatty acids (rTFA), either trans-vaccenic acid (tVA) or trans-palmitoleic acid (tPA), whey protein hydrolysate, leucine or combinations of rTFA and dairy protein compounds. Industrial TFA elaidic acid (EA) was also investigated and compared with rTFA. Inflammatory prostaglandins (PG) and F
Sigma-Aldrich
Methyl formate, reagent grade, 97%
Pricing and availability is not currently available.