Merck
  • Relevance of two genes in the multidrug resistance of hepatocellular carcinoma: in vivo and clinical studies.
Relevance of two genes in the multidrug resistance of hepatocellular carcinoma: in vivo and clinical studies.
Tumori (2010-05-05)
Zhengang Sun, Zizhuo Zhao, Gaopeng Li, Shengli Dong, Zhiyong Huang, Lu Ye, Huifang Liang, Junyuan Qu, Xi Ai, Wanguang Zhang, Xiaoping Chen
ABSTRACT
A former study evaluated the roles of four multidrug resistance-related proteins, namely multidrug resistance protein 1 (MDR1), breast cancer resistance protein (BCRP), multidrug resistance-related protein (MRP1), and lung resistance-related protein (LRP), in the MDR mechanism of the multidrug resistant hepatoma HepG2/ADM cell line and proposed that up-regulated MDR1 and BCRP are responsible for the MDR of hepatocellular carcinoma. This work aims to confirm that assumption in vivo and in clinical specimens. First, the chemotherapeutic resistance of subcutaneous HepG2/ADM tumor and hepatocellular carcinoma samples post-transarterial chemoembolization (TACE) was determined by MTT, contrary to subcutaneous HepG2 tumor and hepatocellular carcinoma samples without TACE, respectively. Then, the mRNA and protein differential expression of the four genes between the MDR tissues and drug-sensitive tissues were quantitatively investigated by real-time RT-PCR and enhanced chemiluminescence western blot analysis, respectively. 1) mRNA expression of BCRP and MDR1 was respectively amplified 38.3 and 20.1 fold in tumors of HepG2/ADM mice compared to those of HepG2 mice, whereas they were respectively augmented for 14.6 and 9.3 times in TACE samples, contrary to the tumor tissues without TACE. 2) The protein presence of MDR1 and BCRP in MDR tumors was also significantly higher than those in the control group in vivo and in clinical specimens. 3) The mRNA expressions of MDR1 and BCRP were correlated to their protein levels. The study showed that MDR1 and BCRP may be the most important factors for drug resistance in hepatocellular carcinoma. Moreover, the positive correlation between their mRNA and protein expression indicates the easy prediction of HCC MDR and possible inhibitive target of drug resistance at multi-levels.