Merck
  • Biological responses and histological studies of estriol and estriol sulfate in fetal and newborn uteri of guinea pig.
Biological responses and histological studies of estriol and estriol sulfate in fetal and newborn uteri of guinea pig.
Journal of steroid biochemistry (1984-04-01)
J R Pasqualini, B L Nguyen
ABSTRACT
The biological responses of estriol (E3) and of estriol-3-sulfate (E3-S) in the fetal and newborn uteri of guinea pig were studied. After a treatment of E3 (1 mg/kg/day) or E3-S (1.4 mg/kg/day) to pregnant guinea pigs (49-60 days of gestation) for 6 days, both estrogens provoke a significant uterotrophic effect in the fetal uterus which increases in weight 1.8-2.5 times in relation to the non-treated animals. The stimulation of progesterone receptor (PR) is also very intense, 7-12 times in relation to the control animals. In another series of experiments newborn guinea pigs (2-day old) were treated with 100 micrograms/animal of E3 or 140 micrograms/animal of E3-S for a short (2 days) or a long (12 days) period. Concerning the uterotrophic effect, the weight of the uterus increases 1.8-2.5 times (in relation to the non-treated animals) after a 2-day treatment and the effect continues to increase up to 4-5 times in the 12-day treated animals. In opposition to the fetal uterus, the effect on PR provoked by E3 or E3-S is very limited (only an increase of 1 time in relation to the non-treated animals); the effect is even less intense after the 12-day treatment. Histological studies show an intense hypertrophic effect particularly in the epithelium of the endometrium in the fetal and newborn uteri for both E3 and E3-S. In the newborns the effect is also intense on the epithelium of the uterine gland. It is concluded that: (1) Estriol and estriol sulfate are very active and with similar intensity on the uterine weight before and after birth; (2) The stimulatory effect on PR decreases very significantly after birth and after a long treatment; (3) E3-S can act as a potent hormonal precursor.
Sigma-Aldrich
Estriol 3-sulfate sodium salt, ≥98% (TLC)