Merck
  • Effect of poly(vinyl acetate-acrylamide) microspheres properties and steric hindrance on the immobilization of Candida rugosa lipase.
Effect of poly(vinyl acetate-acrylamide) microspheres properties and steric hindrance on the immobilization of Candida rugosa lipase.
Bioresource technology (2012-09-20)
Dong-Hao Zhang, Li-Xia Yuwen, Chao Li, Ya-Qiong Li
ABSTRACT
Poly(vinyl acetate-acrylamide) microspheres were synthesized in the absence or presence of isooctane via suspension polymerization and utilized as carriers to immobilize Candida rugosa lipase. When the hydrophobic/hydrophilic surface characteristics of the microspheres were modified by changing the ratio of vinyl acetate (hydrophobic monomer) to acrylamide (hydrophilic monomer) from 50:50 to 86:24, the immobilization ratio changed from 45% to 92% and the activity of the immobilized lipase increased from 202.5 to 598.0 U/g microsphere. Excessive lipase loading caused intermolecular steric hindrance, which resulted in a decline in lipase activity. The maximum specific activity of the immobilized lipase (4.65 U/mg lipase) was higher than that of free lipase (3.00 U/mg lipase), indicating a high activity recovery during immobilization.
Sigma-Aldrich
Poly(vinyl acetate), average Mw ~500,000 by GPC
Sigma-Aldrich
Poly(vinyl acetate), average Mw ~100,000 by GPC, beads
Poly(vinyl acetate) dispersion 30 per cent, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Kollicoat® SR 30 D, 28.5-31.5% solids basis