Merck
  • Study protocol for a non-inferiority trial of cytisine versus nicotine replacement therapy in people motivated to stop smoking.
Study protocol for a non-inferiority trial of cytisine versus nicotine replacement therapy in people motivated to stop smoking.
BMC public health (2011-11-23)
Natalie Walker, Colin Howe, Chris Bullen, Hayden McRobbie, Marewa Glover, Varsha Parag, Jonathan Williman, Reon Veale, Vili Nosa, Joanne Barnes
ABSTRACT
Smokers need effective support to maximise the chances of successful quit attempts. Current smoking cessation medications, such as nicotine replacement therapy (NRT), bupropion, nortriptyline or varenicline, have been shown to be effective in clinical trials but are underused by smokers attempting to quit due to adverse effects, contraindications, low acceptability and/or high cost. Cytisine is a low-cost, plant-based alkaloid that has been sold as a smoking cessation aid in Eastern Europe for 50 years. A systematic review of trial evidence suggests that cytisine has a positive impact on both short- and long-term abstinence rates compared to placebo. However, the quality of the evidence is poor and insufficient for licensing purposes in many Western countries. A large, well-conducted placebo-controlled trial (n = 740) of cytisine for smoking cessation has recently been published and confirms the findings of earlier studies, with 12-month continuous abstinence rates of 8.4% in the cytisine group compared to 2.4% in the placebo group (Relative risk = 3.4, 95% confidence intervals 1.7-7.1). No research has yet been undertaken to determine the effectiveness of cytisine relative to that of NRT. A single-blind, randomised controlled, non-inferiority trial has been designed to determine whether cytisine is at least as effective as NRT in assisting smokers to remain abstinent for at least one month. Participants (n = 1,310) will be recruited through the national telephone-based Quitline service in New Zealand and randomised to receive a standard 25-day course of cytisine tablets (Tabex®) or usual care (eight weeks of NRT patch and/or gum or lozenge). Participants in both study arms will also receive a behavioural support programme comprising an average of three follow-up telephone calls delivered over an eight-week period by Quitline. The primary outcome is continuous abstinence from smoking at one month, defined as not smoking more than five cigarettes since quit date. Outcome data will also be collected at one week, two months and six months post-quit date. Cytisine appears to be effective compared with placebo, and given its (current) relative low cost may be an acceptable smoking cessation treatment for smokers, particularly those in low- and middle-income countries. Cytisine's 'natural' product status may also increase its acceptability and use among certain groups of smokers, such as indigenous people, smokers in countries where the use of natural medicines is widespread (e.g. China, India), and in those people who do not want to use NRT or anti-depressants to help them quit smoking. However it is important to ascertain the effectiveness of cytisine compared with that of existing cessation treatments. Australian New Zealand Clinical Trials Registry (ACTRN12610000590066).
Sigma-Aldrich
Cytisine, ≥99.0% (HPLC), ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Cytisine, ≥99%, powder
Pricing and availability is not currently available.