Merck
  • Synthesis and photophysical properties of complexes of Be(II) and Zn(II) with 10-hydroxybenzo[h]quinoline ligand.
Synthesis and photophysical properties of complexes of Be(II) and Zn(II) with 10-hydroxybenzo[h]quinoline ligand.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-02-28)
Zhiqiang Guo, Zhenming Dong, Ruitao Zhu, Shuo Jin, Bo Liu
ABSTRACT
Two 10-hydroxybenzo[h]quinoline metal complexes, bis(10-hydroxybenzo[h]quinolinato) beryllium (Bebq2) and bis(10-hydroxybenzo[h]quinolinato)zincum (Znbq2), have been synthesized. The structure are characterized by 1HNMR, IR and so on. The photophysical processes of Bebq2 and Znbq2 have been carefully investigated by fluorescence spectra. The results show that the compounds emit yellow-green and yellow light. The emission peaks are at 492 and 512 nm, respectively. In addition, the light-emitting can be quenched by electron donor, N,N-dimethylaniline (DMA), and the quenching process follows the Stern-Volmer equation. Furthermore, the molecular interactions of Bebq2 and Znbq2 with electron acceptor, dimethylterephthalate (DMTP), were also carefully investigated. It displayed that the Znbq2 is a potential substitute for Bebq2 as an excellent emitting material.
Sigma-Aldrich
Dimethyl terephthalate, purum, ≥99.0% (GC)
Supelco
Dimethyl terephthalate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dimethyl terephthalate, ReagentPlus®, ≥99%