Merck
  • Diastereoselective solid-phase radical addition to oxime ether anchored to polymer support.
Diastereoselective solid-phase radical addition to oxime ether anchored to polymer support.
Chemical & pharmaceutical bulletin (2003-05-09)
Hideto Miyabe, Chihiro Konishi, Takeaki Naito
ABSTRACT
Stereocontrol in radical reactions of oxime ether anchored to polymer support was studied. Highly diastereoselective solid-phase radical reaction was achieved by using triethylborane and diethylzinc as a radical initiator at low reaction temperature, providing a novel method for the synthesis of the alpha-amino acid derivatives with excellent diastereoselectivities.
Sigma-Aldrich
Triethylborane solution, 1.0 M in THF
Sigma-Aldrich
Triethylborane solution, 1.0 M in hexanes
Sigma-Aldrich
Triethylborane, ≥95%