Merck
  • Ganoderma lucidum polysaccharide inhibits UVB-induced melanogenesis by antagonizing cAMP/PKA and ROS/MAPK signaling pathways.
Ganoderma lucidum polysaccharide inhibits UVB-induced melanogenesis by antagonizing cAMP/PKA and ROS/MAPK signaling pathways.
Journal of cellular physiology (2018-10-27)
Shuanghai Hu, Jinhua Huang, Shiyao Pei, Yujie Ouyang, Yufang Ding, Ling Jiang, Jianyun Lu, Liyang Kang, Lihua Huang, Hong Xiang, Rong Xiao, Qinghai Zeng, Jing Chen
ABSTRACT
Ultraviolet (UV)-induced pigmentation is very common in clinical practice, but the current treatments are rarely effective, accompanied by some side effects. Ganoderma lucidum polysaccharide (GLP) is a natural antioxidant with no toxic side effects, which can antagonize UVB-induced fibroblast photo aging. The study aims to explore the role of GLP in inhibiting UVB-induced melanogenesis and its possible mechanism. The expression of melanogenesis genes such as microphthalmia-associated transcription factor (MITF), tyrosine (TYR), tyrosinase related protein 1 (TYRP1), tyrosinase related protein 2 (TYRP2), ras-related protein Rab-27A (Rab27A), and Myosin shows an upward trend after exposure of B16F10 and PIG1 cells to UVB irradiation, but GLP can downregulate the expression of genes related to UVB-induced melanogenesis. GLP can inhibit UVB-activated protein kinase A (PKA) and mitogen-activated protein kinase (MAPK) signaling pathways. Besides, GLP protects mitochondria from UVB damage and inhibits reactive oxygen species (ROS) production. Also, UVB-induced cyclic adenosine monophosphate (cAMP) can be inhibited. It has been found in the experiments of UVB-induced skin pigmentation in zebrafish that GLP is capable of inhibiting UVB-induced skin pigmentation. Meanwhile, it can greatly relieve erythema reaction in guinea pig skin caused by high-dosage UVB irradiation. In conclusion, this study shows that GLP can inhibit UVB-induced melanogenesis by antagonizing cAMP/PKA and ROS/MAPK signaling pathways and is a potential natural safe whitening sunscreen additive.