Merck
  • Understanding the electric and nonelectric field components of the cation effect on the electrochemical CO reduction reaction.
Understanding the electric and nonelectric field components of the cation effect on the electrochemical CO reduction reaction.
Science advances (2020-11-08)
A S Malkani, J Li, N J Oliveira, M He, X Chang, B Xu, Q Lu
ABSTRACT
Electrolyte cations affect the activity of surface-mediated electrocatalytic reactions; however, understanding the modes of interaction between cations and reaction intermediates remains lacking. We show that larger alkali metal cations (excluding the thickness of the hydration shell) promote the electrochemical CO reduction reaction on polycrystalline Cu surfaces in alkaline electrolytes. Combined reactivity and in situ surface-enhanced spectroscopic investigations show that changes to the interfacial electric field strength cannot solely explain the reactivity trend with cation size, suggesting the presence of a nonelectric field strength component in the cation effect. Spectroscopic investigations with cation chelating agents and organic molecules show that the electric and nonelectric field components of the cation effect could be affected by both cation identity and composition of the electrochemical interface. The interdependent nature of interfacial species indicates that the cation effect should be considered an integral part of the broader effect of composition and structure of the electrochemical interface on electrode-mediated reactions.
Sigma-Aldrich
Rubidium hydroxide hydrate
Sigma-Aldrich
Cesium hydroxide monohydrate, 99.95% trace metals basis