Merck
  • Tumor Microenvironment-Derived NRG1 Promotes Antiandrogen Resistance in Prostate Cancer.
Tumor Microenvironment-Derived NRG1 Promotes Antiandrogen Resistance in Prostate Cancer.
Cancer cell (2020-07-18)
Zeda Zhang, Wouter R Karthaus, Young Sun Lee, Vianne R Gao, Chao Wu, Joshua W Russo, Menghan Liu, Jose Mauricio Mota, Wassim Abida, Eliot Linton, Eugine Lee, Spencer D Barnes, Hsuan-An Chen, Ninghui Mao, John Wongvipat, Danielle Choi, Xiaoping Chen, Huiyong Zhao, Katia Manova-Todorova, Elisa de Stanchina, Mary-Ellen Taplin, Steven P Balk, Dana E Rathkopf, Anuradha Gopalan, Brett S Carver, Ping Mu, Xuejun Jiang, Philip A Watson, Charles L Sawyers
ABSTRACT
Despite the development of second-generation antiandrogens, acquired resistance to hormone therapy remains a major challenge in treating advanced prostate cancer. We find that cancer-associated fibroblasts (CAFs) can promote antiandrogen resistance in mouse models and in prostate organoid cultures. We identify neuregulin 1 (NRG1) in CAF supernatant, which promotes resistance in tumor cells through activation of HER3. Pharmacological blockade of the NRG1/HER3 axis using clinical-grade blocking antibodies re-sensitizes tumors to hormone deprivation in vitro and in vivo. Furthermore, patients with castration-resistant prostate cancer with increased tumor NRG1 activity have an inferior response to second-generation antiandrogen therapy. This work reveals a paracrine mechanism of antiandrogen resistance in prostate cancer amenable to clinical testing using available targeted therapies.
Product Description
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture