
(R)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride

Synonym(s):
(R)-(−)-MTPA-Cl, Mosher’s acid chloride
Linear Formula:
C6H5C(OCH3)(CF3)COCl
CAS Number:
Molecular Weight:
252.62
Products
Brand
Boiling Point (°C)
Formula Weight
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing