
Pramipexole Related Compound D

Synonym(s):
Dexpramipexole dihydrochloride, (R)-2-Amino-4,5,6,7-tetrahydro-6-(propylamino)benzothiazole dihydrochloride // (R)-Pramipexole dihydrochloride, (R)-4,5,6,7-Tetrahydro-6-(propylamino)-benzothiazole-2-amine dihydrochloride, (R)-Pramipexole dihydrochloride, KNS 760704, R-(+)-Pramipexole dihydrochloride, RPPX, SND 919CL2X
Empirical Formula (Hill Notation):
C10H17N3S · 2HCl
CAS Number:
Molecular Weight:
284.25
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing