
PNGase F from Elizabethkingia meningoseptica

Synonym(s):
N-Glycosidase F, PNGase F from Chryseobacterium meningosepticum, PNGase F from Flavobacterium meningosepticum, Peptide N-glycosidase, PNGase F from Elizabethkingia meningoseptica
CAS Number:
Products
Product Category
Brand
Biological Source
pH Value
Physical Form
Product Line
Purity
Recombinant Host
Quality Segment
Package Size
Showing 1-4 of 4
Compare
Product Number
Description
Pricing