
Picolinafen

Synonym(s):
N-(4-Fluorophenyl)-6-(α,α,α-trifluoro-m-tolyloxy)picolinamide, N-(4-Fluorophenyl)-6-[3-(trifluoromethyl)phenoxy]-2-pyridinecarboxamide
Empirical Formula (Hill Notation):
C19H12F4N2O2
CAS Number:
Molecular Weight:
376.30
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2