
Oxybutynin impurity A

Synonym(s):
4-(Diethylamino)but-2-ynyl (RS)-2-(cyclohex-3-enyl)-2-cyclohexyl-2-hydroxyacetate
Empirical Formula (Hill Notation):
C22H35NO3
Molecular Weight:
361.52
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing