Nabumetone
Synonym(s):
4-(6-Methoxy-2-naphthyl)-2-butanone
Empirical Formula (Hill Notation):
C15H16O2
CAS Number:
Molecular Weight:
228.29
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique