
Metamizole sodium hydrate

Synonym(s):
(antipyrinylmethylamino)methanesulfonic acid sodium salt, 1-Phenyl-2,3-dimethyl-5-pyrazolone-4-methylaminomethanesulfonate sodium, 4-Methylamino-1,5-dimethyl-2-phenyl-3-pyrazolone sodium methanesulfonate, Analgin, Dipyrone, Metamizol, Methampyrone, Methylmelubrin, Noraminopyrine methanesulfonate sodium, Sodium methylaminoantipyrine methanesulfonate, Sulpyrine, [(2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)methylamino]methanesulfonic acid sodium salt
Empirical Formula (Hill Notation):
C13H16N3O4S · Na · H2O
CAS Number:
Molecular Weight:
351.35
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing