
Leflunomide impurity A

Synonym(s):
4-(Trifluoromethyl)aniline, α,α,α-Trifluoro-p-toluidine, 4-Aminobenzotrifluoride
Linear Formula:
CF3C6H4NH2
CAS Number:
Molecular Weight:
161.12
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing