
Hydrogen Peroxide 200 vol

Synonym(s):
Hydrogen peroxide solution
Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Products
Formula Weight
Product Line
Markush Class
Markush Group
Package Size
Showing 1-1 of 1
Compare
Product Number
Description
Pricing