
Eupatilin

Synonym(s):
2-(3,4-Dimethoxyphenyl)-5,7-dihydroxy-6-methoxy-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3′,4′,6-trimethoxyflavone, NSC 122413
Empirical Formula (Hill Notation):
C18H16O7
CAS Number:
Molecular Weight:
344.32
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing