Skip to Content
Merck
All Photos(1)

Key Documents

K4643

Sigma-Aldrich

PKCθ, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC126514, MGC141919, PRKCQ, PRKCT, nPKC-theta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

673-911 nmol/min·mg

mol wt

~110 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRKCQ(5588)

General description

PKCθ (protein kinase C θ) is a serine/threonine kinase, belonging to protein kinase C family.

Biochem/physiol Actions

PKCθ (protein kinase C θ) is required for T cell activation and proliferation. In presence of TCR (T cell receptor) stimulation, it is responsible for activation of NF-AT (nuclear factor of activated T-cells), NF-κB (nuclear factor κ B) and AP1 (adaptor-related protein complex 1). It might be a target for the treatment of type 2 diabetes. PKCθ is needed for fat-mediated insulin resistance in skeletal muscle.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santhakumar Manicassamy et al.
Journal of immunology (Baltimore, Md. : 1950), 178(1), 312-319 (2006-12-22)
A functional immune system not only requires rapid expansion of antigenic specific T cells, but also requires efficient deletion of clonally expanded T cells to avoid accumulation of T cells. Fas/Fas ligand (FasL)-mediated apoptosis plays a critical role in the
J Rennecke et al.
European journal of biochemistry, 242(2), 428-432 (1996-12-01)
13 murine tissues and 12 cell lines were tested for the expression of the novel protein kinase C (PKC) isoenzyme mu. Using two different PKC mu antibodies (sc-639 and P26720), PKC mu was detected in all tissues and cells and
Jason K Kim et al.
The Journal of clinical investigation, 114(6), 823-827 (2004-09-17)
Insulin resistance plays a primary role in the development of type 2 diabetes and may be related to alterations in fat metabolism. Recent studies have suggested that local accumulation of fat metabolites inside skeletal muscle may activate a serine kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service