EGFR
Epidermal growth factor receptor (EGFR), its family members Her-2/ErbB-2, Her-3, Her-4 and their ligands, are involved in over 70% of all cancers. EGFR itself has been implicated in ~30% of all solid human tumors. EGFR is associated not only with the proliferation of tumor cells, but also with enhanced tumor cell survival, angiogenesis and metastatic spread. The enhanced activity of the EGFR due to over-expression, co-expression of the receptor and its ligands, as well as activating mutations is the hallmark of many human carcinomas.
The co-expression of the EGFR and its ligands, especially TGFα and EGF, plays a key role in EGFR-mediated tumorigenesis. EGFR expression is a prognostic indicator, predicting poor survival and indicating an advanced state of the disease. When EGFR is co-expressed with other members of the Her family, the various combinations of Her dimers confer different degrees of malignancy. It has been noted that the co-expression of EGFR with Her-2 and Her-3 is associated with more aggressive clinical behavior. In many types of tumor, including lung, breast, prostate, ovary, gastrointestinal tract and brain, the EGFR receptor is expressed approximately 100 times the normal number of EGF receptors found on the surface of normal cells. Furthermore, expression of high levels of these two receptors in nonmalignant cell lines, either alone or in combination, leads to a transformed phenotype.
Due to all these observations, it is no surprise that EGFR and Her-2/Erb-2 were identified early on as important targets for drug development. Indeed, the first signal transduction therapeutic agent introduced into the clinic was Herceptin, an anti-Her-2 antibody, followed closely by the protein tyrosine kinase inhibitors Iressa (ZD 1839) and Tarceva (OSI-774) and the anti-EGFR antibody Erbitux (mAb 225).
The enhanced activity of the EGFR is due to a number of molecular events. Most common is the overexpression of the receptor along with the expression of the EGFR receptor ligands like TGFα, EGF, amphiregulin and HB-EGF, leading to persistent autocrine stimulation. Another common occurrence is an activating mutation resulting from deletion of exons 2 through 7, leading to a persistently active receptor Δ (2-7)EGFR (also known as EFFRvIII) in the absence of a ligand. The emergence of this mutation occurs in the most aggressive forms of EGFR overexpressing tumors.
Activation of the EGFR pathway is not limited to members of the EGFR family, but frequently occurs due to the transactivation by other signaling pathways such as mitogenic G protein-coupled receptors and the PDGF receptor. Furthermore, the EGFR pathway cooperates in a synergistic manner with pp60c-Src, and the deletion of PTEN, the negative regulator of PKB/Akt. The frequent involvement of EGFR in human tumors has identified it as a target for novel therapies. The first breakthrough was the development of selective EGFR kinase inhibitors (tyrphostins) like tyrphostins, AG 1478 and ZD 1839 (Iressa). Iressa is one of the two kinase inhibitors (the other being Gleevec) to receive approval for clinical application. It is important to note that it was recently found that the response of patients suffering from non-small-cell lung carcinoma to Iressa, is limited to the 7-10% harboring mutations in the kinase domain of the receptor. Other inhibitors, similar to Iressa, like Tarceva (OSI-774), the pan-Her reversible inhibitor GW 2016, the irreversible inhibitor CI-1033, which targets both RGFR and Her-2, are in the pipeline. Antibodies to the EGFR, like Erbitux and TGFα fused to a mutated form of pseudomonas exotoxin, TP-38, have also undergone clinical development.
The Table below contains accepted modulators and additional information.
Family Members | EGFR | ErbB2 |
---|---|---|
Other Names | ErbB-1 (avian erythroblastic leukemia viral (v-erb-b) oncogene homolog) HER1 ERBB crbB | Neu (rat) HER2 |
Molecular Weight | ~180 kDa | ~185 kDa |
Structural Data | 1210 aa | 1255 aa |
Isoforms | Four alternatively spliced transcripts Secreted extracellular domain Auto-activating deletions in the extracellular domain | Two alternatively spliced transcripts Secreted form (Herstatin) |
Species | All four receptors are expressed in mammals. A single ortholog of the receptor is expressed in D. melanogaster and C. elegans | |
Domain Organization | All four receptors have similar structural domains comprising of an extracellular ligand binding domain, a single transmembrane domain, an intracellular tyrosine kinase domain and a large unstructured tail | |
Phosphorylation Sites | Tyr845 Tyr891 Tyr920 Tyr974 Tyr992 Tyr1045 Tyr1068 Tyr1045 Tyr1086 Tyr1101 Tyr1114 Tyr1148 Tyr1173 Tyr654 Tyr669 Ser1046 Ser1047 | Tyr882 Tyr 899 Tyr958 Tyr1023 Tyr1028 Tyr1139 Tyr1143 Tyr1196 Tyr1221/22 Tyr1226 Tyr1227 Tyr1249 Tyr1253 |
Tissue Distribution | Brain, neurons, skeletal muscle, prostate, liver, pancreas, lung, tongue, skin, kidney, trachea | Brain, spinal cord, placenta, prostate, heart, liver, lung, kidney, pancreas |
Subcellular Localization | Not Known | Not Known |
Binding Partners/ Associated Proteins | EGF (E9644) | Neuregulin-1 |
Upstream Activators | Epidermal growth factor (EGF) (E9644) Transforming growth factor-α (TGF-α) (T7924) Amphiregulin (AR) (A7080) Heparin binding-EGF (HB-EGF) (SRP3052) Betacellulin (BTC) (B3670) Epiregulin (EPR) (E8780) Epigen (EPG) (SRP4969) | Does not bind any of the known EGF like ligands |
Downstream Activation | Grb-2-SOS Shc Shp1 c-Src Gab1 PLC-γ PKC c-Cbl | Grb-2-SOS Shc |
Activators | Not Known | Not Known |
Inhibitors | Gefitinib Erlotinib EKB-569 GW572016 PKI-166 AEE-788 CI-1033 (C7249) AG1478 (T4182) | TAK-165 GW572016 AEE-788 CI-1033 (C7249) PKI-166 |
Selective Activators | Not Known | Not Known |
Physiological Function | Receptor for EGF; involved in control of cell growth and differentiation | Essential component of a neuregulin-receptor complex |
Disease Relevance | Glioblastoma, malignant neoplasms and carcinomas including adenocarcinomas of the breast, lung, prostate, pancreas, head and neck, colon, ovary, bladder | Hyperplasias, benign and malignant, neoplasms and carcinomas, including adenocarcinomas of the breast, prostate, lung, stomach, bladder, colon, cervix |
Family Members | ErbB3 | ErbB4 |
---|---|---|
Other Names | HER3 | HER4 |
Molecular Weight | ~190 kDa | ~180 kDa |
Structural Data | 1342 aa | 1308 aa |
Isoforms | Two alternatively spliced transcripts | Two alternatively spliced transcripts called HER4 JM-α and HER4 JM-β |
Species | All four receptors are expressed in mammals. A single ortholog of the receptor is expressed in D. melanogaster and C. elegans | |
Domain Organization | All four receptors have similar structural domains comprising of an extracellular ligand binding domain, a single transmembrane domain, an intracellular tyrosine kinase domain and a large unstructured tail | |
Phosphorylation Sites | Has an impaired kinase and cannot autophosphorylate Tyr1035 Tyr1178 Tyr1180 Tyr1203/5 Tyr1241 Tyr1243 Tyr1257 Tyr1270 Tyr1309 | Tyr1066 Tyr1162 Tyr1066 Tyr1188 Tyr1189 Tyr1242 Tyr1258 Tyr1284 |
Tissue Distribution | Brain, prostate, dorsal root ganglion, liver, placenta, salivary gland, spinal cord, uterus, heart, lung, muscle, pituitary, thyroid, pancreas, kidney | Brain, bone marrow, spinal cord, dorsal root ganglion, testis, liver, skeletal muscle, cardiac myocytes, salivary gland, tongue, skin, trachea, pancreas |
Subcellular Localization | Not Known | Not Known |
Binding Partners/ Associated Proteins | Neuregulin Ebp1 SH2 domain of p85 | Neuregulin-1 β-cellulin |
Upstream Activators | α and β isoforms of Heregulin-1/Neuregulin-1 (HRG/NRG-1α, HRG/NRG-1β) α and β isoforms of Heregulin-2/ Neuregulin-2 (HRG/NRG-2α, HRG/NRG-2β) | Betacellulin (BTC) (B3670) Heparin binding- EGF (HB-EGF) (SRP3052) Epiregulin (EPR) (E8780) Neuregulin-3 (NRG-3) Neuregulin-4 (NRG-4) |
Downstream Activation | Grb-2/7-SOS Shc PI3K (P8615) | Shc Grb-2-SOS PI3K (P8615) |
Activators | Not Known | Not Known |
Inhibitors | Not Known | AEE-788 CI-1033 (C7249) |
Selective Activators | Not Known | Not Known |
Physiological Function | Involved in development of variety of tissues | Interacts with neuregulins NRG-2 NRG-3 Heparin-binding EGF-like growth factor |
Disease Relevance | Malignant neoplasms of the breast, ovary, pancreas, lung, prostate, bladder, colon | Role of ErbB4 in malignancies is not well established. However, it has been implicated in some tumors of the breast, prostate, ovary, brain, lung |
Network error: Failed to fetch