Merck
HomeMedia PreparationAlbumin from Bovine Serum
Albumin from Bovine Serum Frequently Asked Questions (FAQs)
Albumins are the most abundant protein in serum or plasma, accounting for 60% of the total protein content and have an important role in modulating osmotic pressure (fluid shifts between the vascular and tissue compartments). Unlike globulins, albumins have comparatively low molecular weights, are soluble in water, are easily crystallized, and contain an excess of acidic amino acids.
Albumins are soluble in water and can act as a transporter or carrier for many hormones and drugs that are not readily water-soluble. It binds water, Ca2+, Na+, and K+, and due to a hydrophobic cleft, also binds fatty acids, bilirubin, hormones, and drugs. Albumin is used to solubilize lipids and is also used as a blocking agent in western blots or ELISA applications. The proteins also play an important role in regulating pH.
The molecular weight of albumins varies depending on the type of albumin. The mature human albumin consists of 585 amino acids and has a molecular mass of 66 kDa.
The most well-known types of albumin proteins are human, bovine serum albumins and recombinant. Other albumins include chicken, mouse, porcine.
Bovine serum albumin (BSA) is a commonly used small (~66 kDa) globular albumin protein that binds, sequesters, and stabilizes a range of important molecules and proteins. It is broadly used as an additive to cell culture media, especially serum-free media, and provides a range of benefits including protection from oxidative damage and stabilization of other media components such as fatty acids and pyridoxal.
Our BSA products have been used and published in peer-reviewed articles for many applications, including cell culture, IHC, ELISA and many more. We offer a wide variety of BSA products for your research and manufacturing needs.
Albumin is synthesized in the liver by hepatocytes and then is rapidly excreted into the blood stream, with very little albumin stored in the liver. Albumins serve as a modulator of plasma oncotic pressure in human and can transport both endogenous and exogenous ligands, such as drugs. In clinical medicine, human serum albumin is measured as a marker for the nutritional status of an individual patient.
The amino acid sequence and structure of human albumin have been determined. Human albumin is a protein with no carbohydrate content. It is a single polypeptide chain with one free sulfhydryl group on residue # 34 and 17 intrachain disulfide bonds.
BSA contains about 583 amin acid residues and does not have carbohydrates within a single polypeptide chain. It contains 17 interchain disulfide bridges and 1 sulfhydryl group at pH 5-7.
Albumin levels in the blood can be used to diagnose a variety of medical conditions, including liver disease, kidney disease, and malnutrition. Low levels of albumin in the blood can indicate liver or kidney damage, while high levels can indicate dehydration.
Albumins are commonly used in the laboratory as a blocking agent to prevent non-specific binding in immunoassays. They can also be used as a stabilizer in enzyme assays and as a protein standard in protein quantification assays.
  • Western blotting: BSA is commonly used as a blocking agent in Western blotting to prevent non-specific binding of primary and secondary antibodies to the membrane.
  • ELISA (enzyme-linked immunosorbent assay): BSA can be used as a blocking agent in ELISA to prevent non-specific binding of antibodies to the plate.
  • Cell culture: Albumin is commonly used as a supplement in cell culture media to provide cells with essential nutrients and growth factors.
  • Stabilizer: Albumin can be used as a stabilizer in vaccines, pharmaceuticals, and other biological products to protect against degradation.
Albumins are responsible for maintaining the osmotic pressure of the blood by preventing the loss of water from the bloodstream. They do this by attracting water molecules through their polar and charged amino acid residues.
Albumins purified from a variety of methods including the true Cohn fractionation method, heat shock, chromatography, and modified ethanol fractionation methods. Additional purification steps may include charcoal filtration and crystallization.
Purification Methods and Application References for Bovine Serum Albumin
ProductPurification MethodApplication References
A2153Cold-Ethanol
Fractionation
Cell/Tissue Culture: 50,51,52
ELISA Blocking: 74
IHC/ICC/IF: 109, 110
Immunoblotting: 124, 125
Standard/Calibrator: 145
A3156Cold-Ethanol
Fractionation
Cell/Tissue Culture: T
A4378Cold-Ethanol
Fractionation
IHC/ICC/IF: 97
A4503Cold-Ethanol
Fractionation
Cell/Tissue Culture: 26,27,28
ELISA Blocking: 65
IHC/ICC/IF: 98
Immunoblotting: 132, 133, 134
A6003Cold-Ethanol
Fractionation
Cell/Tissue Culture: 46, 47
Standard/Calibrator: 139, 140
Stabilization/Diluent: 10, 155
A8806Cold-Ethanol
Fractionation
Binding, Transport, & Carrier: 4, 5, 6, 7
Cell/Tissue Culture: 48
IHC/ICC/IF: 107
Stabilization/Diluent: 156
A9418Cold-Ethanol
Fractionation
Cell/Tissue Culture: T
A0281Heat-Shock FractionationCell/Tissue Culture: 43, 44, 45
A1470Heat-Shock FractionationBinding, Transport, & Carrier: 1
Cell/Tissue Culture: 1, 16, 17, 18
Immunoblotting: 115
A1595Heat-Shock FractionationCell/Tissue Culture: 53, 54, 55
A2058Heat-Shock FractionationImmunoblotting: 123
Standard/Calibrator: 141, 142, 143
A3059Heat-Shock FractionationCell/Tissue Culture: 30, 56
Immunoblotting: 118, 119
Standard/Calibrator: 137
Stabilization/Diluent: 56, 152
A3294Heat-Shock FractionationChIP/ Sequencing/ Hybridization: 62
IHC/ICC/IF: 108
Standard/Calibrator: 144
A3311Heat-Shock FractionationCell/Tissue Culture: T
A3803Heat-Shock FractionationBinding, Transport, & Carrier: 2, 3
Cell/Tissue Culture: 36
ELISA Blocking: 68
IHC/ICC/IF: 99
Standard/Calibrator: 138
A3858Heat-Shock FractionationELISA Blocking: 5
A3912Heat-Shock FractionationCell/Tissue Culture: 37
IHC/ICC/IF: 100
Immunoblotting: 120
A4161Heat-Shock FractionationCell/Tissue Culture: 23, 24, 25
A4612Heat-Shock FractionationCell/Tissue Culture: 49
A4919Heat-Shock FractionationCell/Tissue Culture: 13, 14
IHC/ICC/IF: 84
A7030Heat-Shock FractionationCell/Tissue Culture: 39, 40, 41, 42
ELISA Blocking: 69, 70, 71, 72, 73
IHC/ICC/IF: 106
Immunoblotting: 122
Stabilization/Diluent: 154
A7888Heat-Shock FractionationCell/Tissue Culture: 31, 32, 33, 34
ELISA Blocking: 67
A7906Heat-Shock FractionationCell/Tissue Culture: 29
ChIP/ Sequencing/ Hybridization: 58, 59, 60
ELISA Blocking: 66
Immunoblotting: 116, 117
Standard/Calibrator: 135, 136
Stabilization/Diluent: 149, 150, 151
A7979Heat-Shock FractionationCell/Tissue Culture: T
A8022Heat-Shock FractionationELISA Blocking: 63
IHC/ICC/IF: 88, 89, 90, 91, 92
Immunoblotting: 92, 112, 113, 114
Stabilization/Diluent: 147
A8412Heat-Shock FractionationCell/Tissue Culture: T
A8577Heat-Shock FractionationStabilization/Diluent: 153
A9085Heat-Shock FractionationCell/Tissue Culture: 15 IHC/ICC/IF: 85, 86, 87
Immunoblotting: 86, 87, 126
A9430Heat-Shock FractionationCell/Tissue Culture: 19
A9543Heat-Shock FractionationCell/Tissue Culture: 35
A9576Heat-Shock FractionationCell/Tissue Culture: T
A9647Heat-Shock Fractionation
IHC/ICC/IF: 76, 77, 78, 79, 80, 81, 82, 83
Immunoblotting: 111
Standard/Calibrator: 127, 128, 129, 130, 131
Stabilization/Diluent: 146
B2064Heat-Shock FractionationCell/Tissue Culture: 38
IHC/ICC/IF: 101, 102, 103, 104
Immunoblotting: 103
B2518Heat-Shock FractionationIHC/ICC/IF: 105
Immunoblotting: 121
B4287Heat-Shock FractionationCell/Tissue Culture: 20, 21, 22
T ELISA Blocking: 64
IHC/ICC/IF: 93, 94, 95, 96
Stabilization/Diluent: 148
B6917Heat-Shock FractionationChIP/ Sequencing/ Hybridization: 57
Table 1.BSA products sorted and cited based on purification method and applications. T = QC test for specified application. ChIP = chromatin immunoprecipitation; ELISA = enzyme-linked immunosorbent assay; IHC = immunohistochemistry; ICC = immunocytochemistry; IF = immunofluorescence.
Albumin is relatively simple to isolate and purify. One of the first methods of isolation involved extensive dialysis of serum against water and removed most globulins. A second procedure took advantage of the good solubility of albumin at low to moderate ammonium sulfate concentrations, and effected precipitation by lowering the pH. Electrophoretic isolation was also employed, as was affinity chromatography. However, none of these methods were applicable to large scale production.
Initial isolation is accomplished by heat treatment or by alcohol precipitation. Most commercial preparations are now prepared by alcohol precipitation, a method developed by E. J. Cohn and his associates in the 1940's ("Fraction V" yields albumin with a purity of about 96%), or by Heat Treatment. The additional removal of impurities can be accomplished by crystallization, preparative electrophoresis, ion exchange chromatography, affinity chromatography (e.g., ConA-agarose removes glycoproteins), heat treatment (removes globulins), low pH treatment, charcoal treatment, organic solvent precipitation (i.e., isooctane), and low temperature treatment. Charcoal treatment and organic solvent precipitation remove fatty acids.
Yes, albumins can be denatured by changes in pH, temperature, and ionic strength. Denaturation can cause the protein to lose its functional properties.
Albumins are readily soluble in water and can only be precipitated by high concentrations of neutral salts such as ammonium sulfate. The solution stability of BSA is very good (especially if the solutions are stored as frozen aliquots). In fact, albumins are frequently used as stabilizers for other solubilized proteins (e.g., labile enzymes). However, albumin is readily coagulated by heat. When heated to 50 °C or above, albumin quite rapidly forms hydrophobic aggregates which do not revert to monomers upon cooling. At somewhat lower temperatures aggregation is also expected to occur, but at relatively slower rates.
pI in Water at 25 °C:Fatty Acid Depleted  - 5.3, Endogenous Material  - 4.7; 4.9
pH of 1% Solution:5.2-7
Optical Rotation:[α] 259 : -61°; [α] 264 : -63°
Stokes Radius (r s ): 3.48 nm
Sedimentation constant, S 20,W X 10 134.5 (monomer), 6.7 (dimer)
Diffusion constant, D 20,W X 10 75.9
Partial specific volume, V 200.733
Intrinsic viscosity, η0.0413
Frictional ratio, f/f 0
1.30
Overall dimensions, Å40 X 140
Refractive index increment1 (578 nm) X 10 -31.90
Optical absorbance, A279 nm (1 gram/liter)0.667
Mean residue rotation1 [ m' ] 2338443
Mean residue ellipticity
21.1 [θ] 209 nm ; 20.1 [θ] 222 nm
Estimated α-helix, %
54
Estimated β-form %
18
Albumins can bind to a variety of molecules, including fatty acids, hormones, and drugs. They do this through hydrophobic and electrostatic interactions.
Related Products
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Lin B, Lin X, Stachel M, Wang E, Luo Y, Lader J, Sun X, Delmar M, Bu L. Culture in Glucose-Depleted Medium Supplemented with Fatty Acid and 3,3?,5-Triiodo-l-Thyronine Facilitates Purification and Maturation of Human Pluripotent Stem Cell-Derived Cardiomyocytes. Front. Endocrinol.. 8 https://doi.org/10.3389/fendo.2017.00253
2.
Boteon YL, Wallace L, Boteon APCS, Mirza DF, Mergental H, Bhogal RH, Afford S. An effective protocol for pharmacological defatting of primary human hepatocytes which is non-toxic to cholangiocytes or intrahepatic endothelial cells. PLoS ONE. 13(7):e0201419. https://doi.org/10.1371/journal.pone.0201419
3.
Aragonès G, Suárez M, Ardid-Ruiz A, Vinaixa M, Rodríguez MA, Correig X, Arola L, Bladé C. 2016. Dietary proanthocyanidins boost hepatic NAD+ metabolism and SIRT1 expression and activity in a dose-dependent manner in healthy rats. Sci Rep. 6(1): https://doi.org/10.1038/srep24977
4.
Liu C, Han T, Stachura DL, Wang H, Vaisman BL, Kim J, Klemke RL, Remaley AT, Rana TM, Traver D, et al. 2018. Lipoprotein lipase regulates hematopoietic stem progenitor cell maintenance through DHA supply. Nat Commun. 9(1): https://doi.org/10.1038/s41467-018-03775-y
5.
Ota A, Kovary KM, Wu OH, Ahrends R, Shen W, Costa MJ, Feldman BJ, Kraemer FB, Teruel MN. 2015. Using SRM-MS to quantify nuclear protein abundance differences between adipose tissue depots of insulin-resistant mice. Journal of Lipid Research. 56(5):1068-1078. https://doi.org/10.1194/jlr.d056317
6.
Patsoukis N, Bardhan K, Chatterjee P, Sari D, Liu B, Bell LN, Karoly ED, Freeman GJ, Petkova V, Seth P, et al. 2015. PD-1 alters T-cell metabolic reprogramming by inhibiting glycolysis and promoting lipolysis and fatty acid oxidation. Nat Commun. 6(1): https://doi.org/10.1038/ncomms7692
7.
Fang L, Xie D, Wu X, Cao H, Su W, Yang J. Involvement of Endoplasmic Reticulum Stress in Albuminuria Induced Inflammasome Activation in Renal Proximal Tubular Cells. PLoS ONE. 8(8):e72344. https://doi.org/10.1371/journal.pone.0072344
8.
Svensson RU, Parker SJ, Eichner LJ, Kolar MJ, Wallace M, Brun SN, Lombardo PS, Van Nostrand JL, Hutchins A, Vera L, et al. 2016. Inhibition of acetyl-CoA carboxylase suppresses fatty acid synthesis and tumor growth of non-small-cell lung cancer in preclinical models. Nat Med. 22(10):1108-1119. https://doi.org/10.1038/nm.4181
9.
Pavani K, Hendrix A, Van Den Broeck W, Couck L, Szymanska K, Lin X, De Koster J, Van Soom A, Leemans B. Isolation and Characterization of Functionally Active Extracellular Vesicles from Culture Medium Conditioned by Bovine Embryos In Vitro. IJMS. 20(1):38. https://doi.org/10.3390/ijms20010038
10.
Heras S, De Coninck DIM, Van Poucke M, Goossens K, Bogado Pascottini O, Van Nieuwerburgh F, Deforce D, De Sutter P, Leroy JLMR, Gutierrez-Adan A, et al. 2016. Suboptimal culture conditions induce more deviations in gene expression in male than female bovine blastocysts. BMC Genomics. 17(1): https://doi.org/10.1186/s12864-016-2393-z
11.
Velasco-Estevez M, Mampay M, Boutin H, Chaney A, Warn P, Sharp A, Burgess E, Moeendarbary E, Dev KK, Sheridan GK. Infection Augments Expression of Mechanosensing Piezo1 Channels in Amyloid Plaque-Reactive Astrocytes. Front. Aging Neurosci.. 10 https://doi.org/10.3389/fnagi.2018.00332
12.
Saka Y, Smith JC. 2007. A mechanism for the sharp transition of morphogen gradient interpretation in Xenopus. BMC Dev Biol. 7(1):47. https://doi.org/10.1186/1471-213x-7-47
13.
Liu H, Chen C, Hung Y, Lin H, Chao H, Shih P, Chuang C, Li W, Huang T, Hsueh Y. RNase A Promotes Proliferation of Neuronal Progenitor Cells via an ERK-Dependent Pathway. Front. Mol. Neurosci.. 11 https://doi.org/10.3389/fnmol.2018.00428
14.
Mao X, Del Bigio MR. 2015. Interference with protease-activated receptor 1 does not reduce damage to subventricular zone cells of immature rodent brain following exposure to blood or blood plasma. J Negat Results BioMed. 14(1): https://doi.org/10.1186/s12952-014-0022-4
15.
Abe K, Zhao L, Periasamy A, Intes X, Barroso M. Non-Invasive In Vivo Imaging of Near Infrared-labeled Transferrin in Breast Cancer Cells and Tumors Using Fluorescence Lifetime FRET. PLoS ONE. 8(11):e80269. https://doi.org/10.1371/journal.pone.0080269
16.
Khraiwesh M, Leed S, Roncal N, Johnson J, Sciotti R, Smith P, Read L, Paris R, Hudson T, Hickman M, et al. 2016. Antileishmanial Activity of Compounds Derived from the Medicines for Malaria Venture Open Access Box Against Intracellular Leishmania major Amastigotes. 94(2):340-347. https://doi.org/10.4269/ajtmh.15-0448
17.
Shekhar A, Lin X, Lin B, Liu F, Zhang J, Khodadadi-Jamayran A, Tsirigos A, Bu L, Fishman GI, Park DS. 2018. ETV1 activates a rapid conduction transcriptional program in rodent and human cardiomyocytes. Sci Rep. 8(1): https://doi.org/10.1038/s41598-018-28239-7
18.
Gustafson-Wagner E, Stipp CS. The CD9/CD81 Tetraspanin Complex and Tetraspanin CD151 Regulate ?3?1 Integrin-Dependent Tumor Cell Behaviors by Overlapping but Distinct Mechanisms. PLoS ONE. 8(4):e61834. https://doi.org/10.1371/journal.pone.0061834
19.
Agrawal S, Guess AJ, Chanley MA, Smoyer WE. 2014. Albumin-induced podocyte injury and protection are associated with regulation of COX-2. Kidney International. 86(6):1150-1160. https://doi.org/10.1038/ki.2014.196
20.
Adhikary G, Grun D, Kerr C, Balasubramanian S, Rorke EA, Vemuri M, Boucher S, Bickenbach JR, Hornyak T, Xu W, et al. Identification of a Population of Epidermal Squamous Cell Carcinoma Cells with Enhanced Potential for Tumor Formation. PLoS ONE. 8(12):e84324. https://doi.org/10.1371/journal.pone.0084324
21.
Tomlinson S, Taylor PW, Morgan BP, Luzio JP. 1989. Killing of gram-negative bacteria by complement. Fractionation of cell membranes after complement C5b-9 deposition on to the surface of Salmonella minnesota Re595. 263(2):505-511. https://doi.org/10.1042/bj2630505
22.
Fisher ML, Ciavattone N, Grun D, Adhikary G, Eckert RL. 2017. Sulforaphane reduces YAP/?Np63? signaling to reduce cancer stem cell survival and tumor formation. Oncotarget. 8(43):73407-73418. https://doi.org/10.18632/oncotarget.20562
23.
?mít D, Fouquet C, Pincet F, Zapotocky M, Trembleau A. Axon tension regulates fasciculation/defasciculation through the control of axon shaft zippering. 6 https://doi.org/10.7554/elife.19907
24.
Yu Y, Blokhuis B, Derks Y, Kumari S, Garssen J, Redegeld F. 2018. Human mast cells promote colon cancer growth via bidirectional crosstalk: studies in 2D and 3D coculture models. OncoImmunology. 7(11):e1504729. https://doi.org/10.1080/2162402x.2018.1504729
25.
Buchser WJ, Slepak TI, Gutierrez?Arenas O, Bixby JL, Lemmon VP. 2010. Kinase/phosphatase overexpression reveals pathways regulating hippocampal neuron morphology. Mol Syst Biol. 6(1):391. https://doi.org/10.1038/msb.2010.52
26.
Schöneberg J, Dambournet D, Liu T, Forster R, Hockemeyer D, Betzig E, Drubin DG. 2018. 4D cell biology: big data image analytics and lattice light-sheet imaging reveal dynamics of clathrin-mediated endocytosis in stem cell?derived intestinal organoids. MBoC. 29(24):2959-2968. https://doi.org/10.1091/mbc.e18-06-0375
27.
Kuboyama K, Tanga N, Suzuki R, Fujikawa A, Noda M. Protamine neutralizes chondroitin sulfate proteoglycan-mediated inhibition of oligodendrocyte differentiation. PLoS ONE. 12(12):e0189164. https://doi.org/10.1371/journal.pone.0189164
28.
Mehta V, Abi-Nader KN, Shangaris P, Shaw SWS, Filippi E, Benjamin E, Boyd M, Peebles DM, Martin J, Zachary I, et al. Local Over-Expression of VEGF-D?N?C in the Uterine Arteries of Pregnant Sheep Results in Long-Term Changes in Uterine Artery Contractility and Angiogenesis. PLoS ONE. 9(6):e100021. https://doi.org/10.1371/journal.pone.0100021
29.
Li L, Ma P, Liu Y, Huang C, O W, Tang F, Zhang JV. Intermedin Attenuates LPS-induced Inflammation in the Rat Testis. PLoS ONE. 8(6):e65278. https://doi.org/10.1371/journal.pone.0065278
30.
Zujur D, Kanke K, Lichtler AC, Hojo H, Chung U, Ohba S. 2017. Three-dimensional system enabling the maintenance and directed differentiation of pluripotent stem cells under defined conditions. Sci. Adv.. 3(5):e1602875. https://doi.org/10.1126/sciadv.1602875