Merck
HomeGas Chromatography (GC)Melamine Analysis via GC-MS
Melamine Analysis via GC-MS
Katherine Stenerson, Michael D. Buchanan
Reporter US Volume 28.3
Site Overview
Introduction
Melamine contamination in food became an issue in recent years after the discovery of it, and related compounds, in pet food and baby formula. It was discovered that melamine was intentionally added to inflate nitrogen content, often the sole measure of the amount of protein in these products. The tainted food led to numerous illnesses, several fatalities, and massive product recalls. Currently, imported raw materials, namely wheat gluten and rice protein used to make these foods, as well as the actual consumer-ready foods, may undergo testing to ensure the absence of these compounds. We have detailed preparation and analytical procedures for these adulterants, using HPLC-MSMS instrumentation in previous publications1,2. In this article, we focus on the analysis of melamine and related compounds with the use of more economical gas chromatography-mass spectrometry (GC-MS) instrumentation.
GC-MS Method
The United States Food and Drug Administration (US FDA) adopted a screening method in October 2008 for the GC-MS analysis of melamine and related compounds in a variety of matrices3. Per the method, 0.5 g of the sample is mixed thoroughly with 20 mL of an extraction solvent mixture (10:40:50 diethylamine:water:acetonitrile). Following sonication (30 minutes) and centrifugation (10 minutes), an aliquot is filtered and evaporated to dryness. Sylon™ BFT and pyridine are then added along with an internal standard. The extract is then incubated (70 °C for 45 minutes) so that trimethylsilyl (TMS) derivatives of each analyte are formed. The resulting derivatized extract is then analyzed by GC-MS. The method allows the operation of the MS in the scan mode (m/z from 50-450 amu) or the selected ion monitoring (SIM) mode. Table 1 shows the structures of the four analytes (melamine, ammeline, ammelide, and cyanuric acid) plus 2,6-diamino-4-chloropyrimidine, the internal standard (I.S.) specified by the method.
The image displays the chemical structures of five compounds. At the top, there are three compounds: Melamine, Ammeline, and Ammelide, each labeled with their respective identifiers (G004438, G005015, G005016). Below them are two additional compounds: Cyanuric acid, labeled G005017, and 2,6-Diamino-4-chloropyrimidine, labeled G005018. Each compound is represented with its molecular formula, showcasing the arrangement of atoms and functional groups.
Table 1.Melamine and Related Compounds
Experimental
For this work, we chose a common dry dog food obtained from a local grocery store. The following samples were prepared:
  1. Three calibration standards, each containing all four analytes, were made at levels of 10 ng/mL, 50 ng/mL, and 100 ng/mL (I.S. added at 1000 ng/mL in each), derivatized, and then used to perform a three-point calibration of the instrument.
  2. A laboratory blank was extracted (I.S. added at 1000 ng/mL), derivatized, and then analyzed to show cleanliness.
  3. A 0.5 g dog food sample was extracted (I.S. added at 1000 ng/ mL), derivatized, and then analyzed to determine analyte levels.
  4. A second 0.5 g dog food sample (spiked with each analyte at 10 μg/g) was extracted (I.S. added at 1000 ng/mL), derivatized, and then analyzed to determine method sensitivity and accuracy.
All standards and extracts were analyzed with the MS operating in the scan mode, and again later with the MS operating in the SIM mode.
Results
The following results were observed:
  • MS Mode: Sensitivity was noticeably better when the MS was operated in the SIM mode. All chromatograms shown are from SIM mode analyses.
  • Instrument Calibration: The 100 ng/mL standard is shown in Figure 1. Note the symmetrical peak shape for each analyte and the I.S., achieved because the activity of amide functional groups was minimized when TMS derivatives were formed, and also due to the inert nature of the capillary GC column.
  • Laboratory Blank (Figure 2): Trace levels of each of the four target analytes were detected in the laboratory blank only when the MS was operated in the SIM mode.
  • Dog Food Sample (Figure 3): The detection of analytes was at a level consistent with that observed in the laboratory blank. We concluded that this dog food was not contaminated with any of the target analytes.
  • Spiked Dog Food Sample (Figure 4): The percent recoveries of each the four target analytes are summarized in Table 2. Good recovery was obtained for each analyte.
column: SLB™-5ms, 30m x 0.25 mm I.D., 0.25 μm (28471-U)
oven: 115 °C (3 min.), 10 °C/min. to 325 °C (6 min.)
inj.: 250 °C
MSD interface: 325 °C
scan range: SIM
carrier gas: helium, 1 mL/min., constant
injection: 1 μL, splitless
liner: 4 mm I.D. single taper
sample: 100 ng/mL standard containing each analyte at 100 ng/mL, then I.S. added at 1000 ng/mL, in pyridine:Sylon™ BFT to form TMS derivatives
  1. Cyanuric acid
  2. Ammelide
  3. 2,6-Diamino-4-chloropyrimidine (I.S.)
  4. Ammeline
  5. Melamine
A chromatogram image displaying the results of a gas chromatography-mass spectrometry (GC-MS) analysis. The setup includes a SLB™-ms column and details such as oven temperature settings, injection volume, and carrier gas specifications. The chromatogram shows peaks corresponding to five analytes: 1. Cyanuric acid, 2. Armmelide, 3. 2,6-Diamino-4-chloropyrimidine (internal standard), 4. Armmelide, and 5. Melamine, plotted against time (minutes) on the x-axis. The peaks are labeled with corresponding numbers.
Figure 1.100 ng/mL Calibration Standard (SIM Mode) (referenced above is 28471-U)
sample: extract of a laboratory blank, then I.S. added at 1000 ng/mL, in pyridine: Sylon BFT to form TMS derivatives
Other conditions the same as Figure 1. See Figure 1 for Peak IDs.
A chromatogram image showing the results of a gas chromatography-mass spectrometry (GC-MS) analysis of an extract from a laboratory blank, with an internal standard added at a concentration of 1000 ng/mL in pyridine:Sylon BFT. The chromatogram displays peaks plotted against time (minutes) on the x-axis, with five peaks labeled with corresponding numbers. The text indicates that other conditions are the same as in a referenced figure for peak identification.
Figure 2.Laboratory Blank (SIM Mode)
sample: extract of dog food, then I.S. added at 1000 ng/mL, in pyridine: Sylon BFT to form TMS derivatives
Other conditions the same as Figure 1. See Figure 1 for Peak IDs.
A chromatogram image displaying the results of a gas chromatography-mass spectrometry (GC-MS) analysis of an extract from dog food, with an internal standard added at a concentration of 1000 ng/mL in pyridine:Sylon BFT. The chromatogram shows peaks plotted against time (minutes) on the x-axis, with five peaks labeled with corresponding numbers. The text notes that other conditions are the same as in a referenced figure for peak identification.
Figure 3.Dog Food Sample (SIM Mode)
sample: dog food spiked with each analyte at 10 µg/g, then extracted, then I.S. added at 1000 ng/mL, in pyridine: Sylon BFT to form TMS derivatives
Other conditions the same as Figure 1. See Figure 1 for Peak IDs.
A chromatogram image illustrating the results of a gas chromatography-mass spectrometry (GC-MS) analysis of dog food spiked with each analyte at a concentration of 10 µg/g, followed by extraction and the addition of an internal standard at 1000 ng/mL in pyridine:Sylon BFT. The chromatogram displays multiple peaks plotted against time (minutes) on the x-axis, with five peaks labeled with corresponding numbers. The text indicates that other conditions are the same as in a referenced figure for peak identification.
Figure 4.Spiked Dog Food Sample (SIM Mode)
Analyte% Recovery
Melamine73
Ammeline77
Ammelide105
Cyanuric acid97
Table 2.Recovery from Dog Food Spiked at 10 μg/g
Conclusions
Our observation is that the method is very easy to perform and provides good sensitivity. In particular, the use of the specified extraction solvent mixture was found to be very effective in solubilizing and extracting all target analytes. Additionally, the formation of TMS derivatives allows these analytes to be analyzed by GC, with symmetrical peak shapes, high signal-to-noise ratios, and low detection levels.
Related Products
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Shimelis O, Santasania C. 2009 . Analysis of Melamine in Milk Analytix 1 6-7. lit. code T409021 LCX.
2.
Shimelis O, Santasania C, Trinh A. 2008 . The Extraction and Analysis of Melamine in Milk-Based Products using Discovery DSC-SCX SPE and Ascentis Express HILIC LC-MS/MS; Sigma-Aldrich Technical Report T408188 LEN . [Internet]. Available from: https://www.sigmaaldrich.com/content/dam/sigma-aldrich/docs/Supelco/Application_Notes/t408188-melamine-analysis.pdf
3.
Litzau JJ, Mercer GE, Mulligan KJ. GC-MS Screen for the Presence of Melamine, Ammeline, Ammelide, and Cyanuric Acid US FDA Laboratory Information Bulletin, LIB No. 4423, Volume 24 https://www.fda.gov/food/laboratory-methods-food/laboratory-information-bulletin-lib-4423-melamine-and-related-compounds. [Internet].[cited 31 Dec 2007].
Trademark Attributes
  • Sylon is a trademark of Sigma-Aldrich Co. LLC
  • SLB is a trademark of Sigma-Aldrich Co. LLC