Merck
HomeNucleic Acid Gel ElectrophoresisPrecast Agarose Gels for RNA Electrophoresis
Precast Agarose Gels for RNA Electrophoresis
Product No: P6222
Product Description
Precautions and Disclaimer
Storage
Product Profile
Reagents Required but Not Provided
Procedure
Related Products
Materials
Product Description
Our Precast Agarose Gels for RNA Electrophoresis are suitable for separating RNA sizes from 0.25 to 10 kb. The 8-well gels are 1.25% RNase-free agarose (A9539) prepared using 1X MOPS buffer with no denaturants. The gels are formulated without formaldehyde and are recommended for the analysis of total RNA, in vitro RNA transcripts and Northern blotting.
Denaturation of the RNA is usually not necessary before loading the gel. However, if significant secondary structure of the RNA is suspected, the RNA should be denatured using one of the protocols included in the procedure section.
Precautions and Disclaimer
Our Precast Agarose Gels for RNA electrophoresis are for laboratory use only. Not for drug, household or other uses. Many reagents used in electrophoresis are hazardous. Warning statements are included on the label. Always wear personal protective equipment including gloves and eye protection when handling. Refer to Safety Data Sheet.
Storage
Store flat at room temperature (18 °C-25 °C). See product label for expiration date. Do Not Freeze.
Product Profile
Our Precast Agarose Gels for RNA analysis are suitable for use with Bio-Rad Mini-Sub®, Hoefer Minnie the Gel-Cicle® , Life Technologies Horizon® 58 and our submarine mini-gel (E0638) electrophoresis units.
Gel Descriptions
Tray dimension: 6.8 cm x 10.2 cm
Gel dimension: 6.0 cm x 9.5 cm 
Gel thickness: 5.5 mm
Sample format: Gels are cast with 8 wells that will each
accommodate up to 15 µl sample volume.
Reagents Required but Not Provided
  • 10X MOPS running buffer, (M5755) (0.4 M MOPS, pH approx. 7.0, 0.1 M sodium acetate, 10 mM EDTA)
  • RNA sample loading buffer, (R4268 - with 50 µg/mL ethidium bromide) or (R1386 - without ethidium bromide); 62.5% (v/v) deionized formamide, 1.14 M formaldehyde, 200 µg/mL bromophenol blue, 200 µg/mL xylene cyanole, 1.25 X MOPS-EDTA-sodium acetate, or
  • Gel loading solution, (G7654) (40% (w/v) sucrose, 0.25% (w/v) bromophenol blue, 0.25% (w/v) xylene cyanole FF)
Additional reagents required for specific denaturation methods:
Formamide (F9037)
Dimethyl sulfoxide (D8418)
Glyoxal (128465)
Mixed Bed Resin (for deionization of formamide or glyoxal) (M8032)
Deionization procedure is included with the resin.
Procedure
Prepare sample using one of the following procedures:
A.    No denaturation required
       Bring RNA up to a volume of 8 µl with RNase-free water or 1X MOPS buffer. Add 2 µl of
       gel loading solution (G7654).
 B.   Formamide-only denaturation
       Bring RNA up to a volume of 8 µl with RNase-free water. Add 2 µl 10X MOPS buffer and 9 µl deionized
       formamide. Mix and heat samples for 10 minutes at 70 °C, then chill on ice for at least one minute.
 C.   Formaldehyde denaturation
       Bring RNA up to a volume of 6 µl with RNase-free water. Add 12 µl RNA sample loading buffer (with or without
       ethidium bromide). Mix and heat samples for 10 minutes at 70 °C, then chill on ice for at least one minute.
 D.   Glyoxal denaturation
       Prepare glyoxal/DMSO mixture by mixing 2.5 mL DMSO with 1.5 mL deionized glyoxal and 1 mL 10X MOPS buffer.
       Quickly dispense small aliquots into microcentrifuge tubes and store at –80 °C. Thaw each aliquot only once; do
       not reuse.
       Bring RNA up to a volume of 9 µl with RNase-free water. Add 9 µl glyoxal/DMSO mixture. Mix and heat samples
       for 60 minutes at 50 °C, then chill on ice for at least one minute.
Our Precast Agarose Gels for RNA analysis require less than 5 minutes to set up.
1.    Peel the paper backing from the adhesive strips on the bottom of the tray.
2.    Peel off the lid. Leave the gel in the tray.
3.    Press the tray directly onto the chamber platform. Align the wells so the RNA samples will run straight.
4.    Pour 1X MOPS running buffer in the chamber to a depth of 5 mm OVER the flange of the tray.
5.    Load the RNA sample (≤15 μl volume).
6.    Electrophorese the gels at 3.5 V/cm for 2 hours. Lower voltages for longer times are acceptable.
7.    Remove the gel from the tray to photograph/document and/or destain.
Related Products
RNaseZAP (R2020)
RNA markers:
    0.28-6.6 kb (R7644)
    0.36-9.5 kb (R7020)
RNA marker template set (R4142)
Materials
Loading
References
1.
Farrell R. 1998. RNA Methodologies: A Laboratory Guide for Isolation and Characterization. 2nd ed.. San Diego, CA: Academic Press.
2.
Krieg P. 1996. A Laboratory Guide to RNA: Isolation, Analysis and Synthesis. New York: Wiley-Liss.
3.
Ausubel F. 1995. Short Protocols in Molecular Biology. 3 ed. New York: John Wiley & Sons.
4.
Sambrook J, Fritsch E, Maniatis T. 1989. Molecular Cloning: A Laboratory Manual Vol. 1, 2, and 3. 2nd ed. Cold Spring Harbor, NY: Cold Spring Harbor Laboratory.