Merck
HPLC Analysis of Warfarin Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of Warfarin Enantiomers on Astec® CHIROBIOTIC® V2 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® V2 Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Application
Product No.
Description
Pricing
HPLC Analysis of Warfarin Enantiomers on Astec® Cellulose DMP
application for HPLC
standard
Product No.
Description
Pricing
Warfarin
analytical standard
Warfarin
PESTANAL®, analytical standard
(R)-(+)-Warfarin
≥97% (HPLC)
(S)-(−)-Warfarin
≥97% (HPLC)
CONDITIONS
column
CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 μm particles (15024AST)
mobile phase
[A] 10 mM ammonium acetate, pH 4.1; [B] acetonitrile; (70:30, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 280 nm
injection
10 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC
null