Merck
HPLC Analysis of Ephedrine Compounds on Ascentis® Express HILIC
HPLC Analysis of Ephedrine Compounds on Ascentis® Express HILIC application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express HILIC, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
CONDITIONS
column
Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
mobile phase
[A] acetonitrile; [B] 40 mM ammonium acetate in water; (90:10, A:B)
flow rate
0.4 mL
column temp.
35 °C
detector
UV, 254 nm
injection
1 μL
sample
150 μg/mL each in mobile phase
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany