GC Analysis of 1-(α-Naphthyl)ethylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ G-DA
![GC Analysis of 1-(α-Naphthyl)ethylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ G-DA suitable for GC](/deepweb/assets/sigmaaldrich/product/images/367/789/ce5b821f-2952-4640-a642-0a3778b3f6c5/800/ce5b821f-2952-4640-a642-0a3778b3f6c5.jpg)
Materials
standard
Product No.
Description
Pricing
CONDITIONS
column
Astec CHIRALDEX G-DA, 30 m x 0.25 mm I.D., 0.12 μm (72033AST)
oven
160 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 35 psi
sample
peak 1: R(+)-1-(α-naphthyl)ethylamine
peak 2: S(-)-1-(α-naphthyl)ethylamine
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC