GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Equity-1, 15 m × 0.10 mm I.D., 0.10 μm (28039-U)
oven
175 °C, 30 °C/min to 275 °C (1 min)
inj. temp.
280 °C
detector
FID, 280 °C
carrier gas
hydrogen, 45 cm/sec constant
injection
0.5 μL, 200:1 split
liner
4 mm I.D., split, cup design
sample
Bacterial Acid Methyl Ester (BAME) Mix (47080-U), methyl ester derivatives, total concentration of 10 mg/mL in methyl caproate
Description
Legal Information
null