Merck
Modification of Ti6Al4V surface by diazonium compounds.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-10-06)
Mariusz Sandomierski, Tomasz Buchwald, Beata Strzemiecka, Adam Voelkel
ABSTRACT
Ti6Al4V alloy is the most commonly used in orthopedic industry as an endoprosthesis. Ti6Al4V exhibits good mechanical properties, except the abrasion resistance. Surface modification of Ti6Al4V in order to obtain organic layer, and then the attachment of the polymer, can allow for overcoming this problem. The aim of the work was the modification of Ti6Al4V surface by diazonium compounds: salt or cation generated in situ and examine the influence of the reducing agent - ascorbic acid, and the temperature of reaction on modification process. Moreover, the simulated body fluid was used for the assessment of the organic layer stability on Ti6Al4V surface. The evaluation of the modification was carried out using the following methods: Raman microspectroscopy, scanning electron microscopy and energy-dispersive X-ray spectroscopy. Higher temperature of modification by 4-hydroxymethylbenzenediazonium cation, provides the largest amount of organic layer on the Ti6Al4V alloy. In the case of the Ti6Al4V modified by Variamine Blue B salt, the amount of organic layer is not dependent on the reaction condition. Moreover, the ascorbic acid and the presence of TiO
Sigma-Aldrich
Magnesium chloride hexahydrate, AR, ≥99%
Sigma-Aldrich
4-Aminobenzyl alcohol, 98%