- Poly(ADP-ribose) polymerase 1 contributes to oxidative stress through downregulation of sirtuin 3 during cisplatin nephrotoxicity.
Poly(ADP-ribose) polymerase 1 contributes to oxidative stress through downregulation of sirtuin 3 during cisplatin nephrotoxicity.
Anatomy & cell biology (2016-10-11)
Sang Pil Yoon, Jinu Kim
PMID27722009
ABSTRACT
Enhanced oxidative stress is a hallmark of cisplatin nephrotoxicity, and inhibition of poly(ADP-ribose) polymerase 1 (PARP1) attenuates oxidative stress during cisplatin nephrotoxicity; however, the precise mechanisms behind its action remain elusive. Here, using an
Sigma-Aldrich
Anti-GPX1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution