Merck
A human monoclonal antibody that binds serotype A botulinum neurotoxin.
Hybridoma (2005) (2008-02-26)
Sharad P Adekar, R Mark Jones, M D Elias, Fetweh H Al-Saleem, Michael J Root, Lance L Simpson, Scott K Dessain
ABSTRACT
Monoclonal antibodies have demonstrated significant potential as therapeutics for botulinum neurotoxin exposures. We previously described a hybridoma method for cloning native human antibodies that uses a murine myeloma cell line that ectopically expresses the human telomerase catalytic subunit gene (hTERT) and the murine interleukin-6 gene (mIL-6). Here we describe a heterohybridoma cell line that ectopically expresses mIL-6 and hTERT and has improved stability of hTERT expression. We fused this cell line to human peripheral blood B cells from a subject who had received the botulinum toxoid vaccine, cloning a high-affinity antibody (13A) specific for serotype A botulinum neurotoxin (BoNT/A). The 13A antibody is an affinity-matured, post-germinal center IgG(1) lambda antibody that has partial neutralization activity in vivo. 13A binds an epitope on BoNT/A that overlaps the binding epitope of an IgG antibody previously shown to fully neutralize a lethal dose of BoNT/A in vivo. The 13A antibody may be useful for diagnostic testing or for incorporation into an oligoclonal therapeutic to counteract BoNT/A exposure.