Merck
  • Recognition of a glycosylation substrate by the O-GlcNAc transferase TPR repeats.
Recognition of a glycosylation substrate by the O-GlcNAc transferase TPR repeats.
Open biology (2017-07-01)
Karim Rafie, Olawale Raimi, Andrew T Ferenbach, Vladimir S Borodkin, Vaibhav Kapuria, Daan M F van Aalten
ABSTRACT
O-linked