Merck
  • TOM1 is a PI5P effector involved in the regulation of endosomal maturation.
TOM1 is a PI5P effector involved in the regulation of endosomal maturation.
Journal of cell science (2015-01-16)
Frédéric Boal, Rana Mansour, Marion Gayral, Estelle Saland, Gaëtan Chicanne, Jean-Marie Xuereb, Marlène Marcellin, Odile Burlet-Schiltz, Philippe J Sansonetti, Bernard Payrastre, Hélène Tronchère
ABSTRACT
Phosphoinositides represent a major class of lipids specifically involved in the organization of signaling cascades, maintenance of the identity of organelles and regulation of multiple intracellular trafficking steps. We previously reported that phosphatidylinositol 5-monophosphate (PI5P), produced by the Shigella flexneri phosphatase IpgD, is implicated in the endosomal sorting of the epidermal growth factor receptor (EGFR). Here, we show that the adaptor protein TOM1 is a new direct binding partner of PI5P. We identify the domain of TOM1 involved in this interaction and characterize the binding motif. Finally, we demonstrate that the recruitment of TOM1 by PI5P on signaling endosomes is responsible for the delay in EGFR degradation and fluid-phase bulk endocytosis. Taken together, our data strongly suggest that PI5P enrichment in signaling endosomes prevents endosomal maturation through the recruitment of TOM1, and point to a new function of PI5P in regulating discrete maturation steps in the endosomal system.
Product Description
Avanti
18:1 PI(5)P, 1,2-dioleoyl-sn-glycero-3-phospho-(1′-myo-inositol-5′-phosphate) (ammonium salt), powder