Merck
  • High efficiency reduction capability for the formation of Fab׳ antibody fragments from F(ab)
High efficiency reduction capability for the formation of Fab׳ antibody fragments from F(ab)
Biochemistry and biophysics reports (2015-04-25)
Victor Crivianu-Gaita, Alexander Romaschin, Michael Thompson
ABSTRACT
Antibodies have widespread applications in areas ranging from therapeutics to chromatography and protein microarrays. Certain applications require only the fragment antigen-binding (Fab) units of the protein. This study compares the cleavage efficacy of dithiothreitol (DTT), mercaptoethylamine (MEA), and dithiobutylamine (DTBA) - a relatively new reducing agent synthesized in 2012. Pseudo-first order kinetic analyses show DTBA to be ~213 times faster than DTT and ~71 times faster than MEA in the formation of Fab׳ antibody fragments from polyclonal rabbit antibodies. Monoclonal mouse antibodies were also used to show the feasibility of the reduction process on antibodies from a different species and with a different clonality. DTBA cleaved the monoclonal mouse F(ab)