Merck
  • Clinical-grade human neural stem cells promote reparative neovascularization in mouse models of hindlimb ischemia.
Clinical-grade human neural stem cells promote reparative neovascularization in mouse models of hindlimb ischemia.
Arteriosclerosis, thrombosis, and vascular biology (2013-11-10)
Rajesh Katare, Paul Stroemer, Caroline Hicks, Lara Stevanato, Sara Patel, Randolph Corteling, Erik Miljan, Indira Vishnubhatla, John Sinden, Paolo Madeddu
ABSTRACT
CTX0E03 (CTX) is a clinical-grade human neural stem cell (hNSC) line that promotes angiogenesis and neurogenesis in a preclinical model of stroke and is now under clinical development for stroke disability. We evaluated the therapeutic activity of intramuscular CTX hNSC implantation in murine models of hindlimb ischemia for potential translation to clinical studies in critical limb ischemia. Immunodeficient (CD-1 Fox(nu/nu)) mice acutely treated with hNSCs had overall significantly increased rates and magnitude of recovery of surface blood flow (laser Doppler), limb muscle perfusion (fluorescent microspheres, P<0.001), and capillary and small arteriole densities in the ischemic limb (fluorescence immunohistochemistry, both P<0.001) when compared with the vehicle-treated group. Hemodynamic and anatomic improvements were dose related and optimal at a minimum dose of 3×10(5) cells. Dose-dependent improvements in blood flow and increased vessel densities by hNSC administration early after ischemia were confirmed in immunocompetent CD-1 and streptozotocin-induced diabetic mice, together with marked reductions in the incidence of necrotic toes (P<0.05). Delayed administration of hNSCs, 7 days after occlusion, produced restorative effects when comparable with acute treatment of 35 days after hindlimb ischemia. Histological studies in hindlimb ischemia immunocompetent mice for the first 7 days after treatment revealed short-term hNSC survival, transient elevation of early host muscle inflammatory, and angiogenic responses and acceleration of myogenesis. hNSC therapy represents a promising treatment option for critical limb ischemia.
Sigma-Aldrich
Anti-GDF5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution